Anti-CD27 Mouse Monoclonal Antibody [clone: 203.6]

Proveedor: Cayman Chemical
CAYM10004837-1EA 443 EUR
CAYM10004837-1
Anti-CD27 Mouse Monoclonal Antibody [clone: 203.6]
Anticuerpos
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: 203.6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR