Anti-DLX2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA329349EA 451 EUR
ORIGTA329349
Anti-DLX2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: DLX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR