Anti-ZFP1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA329343EA 435 EUR
ORIGTA329343
Anti-ZFP1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ZFP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR