Anti-Catsper4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA328777EA 952 EUR
ORIGTA328777
Anti-Catsper4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:2000

Type: Primary
Antigen: Catsper4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR