Anti-Gja5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA328773EA 864 EUR
ORIGTA328773
Anti-Gja5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:2000

Type: Primary
Antigen: Gja5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR