Anti-Trpv4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA328772EA 953 EUR
ORIGTA328772
Anti-Trpv4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:2000; Immunochemistry: 1:100-1:3000

Type: Primary
Antigen: Trpv4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR