Anti-TRPC7 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA328754EA 864 EUR
ORIGTA328754
Anti-TRPC7 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:2000; Flow Cytometry: 1:50-1:600

Type: Primary
Antigen: TRPC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR