Anti-KLK7 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590407EA 684 EUR
ORIGTA590407
Anti-KLK7 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; ELISA: 1:100-1:2000; IHC: 1:600; Immunofluorescence: 1:150

Type: Primary
Antigen: KLK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR