Anti-KLK11 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590401EA 568 EUR
ORIGTA590401
Anti-KLK11 Rabbit Polyclonal Antibody
Recommended Dilutions: ELISA: 1:100-1:2000; IHC: 1:650; Immunofluorescence: 1:162

Type: Primary
Antigen: KLK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR