Anti-B3GNT6 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA329578EA 500 EUR
ORIGTA329578
Anti-B3GNT6 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: B3GNT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR