Anti-TFAM Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA329264EA 575 EUR
ORIGTA329264
Anti-TFAM Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TFAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR