Anti-CCNE2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590293EA 548 EUR
ORIGTA590293
Anti-CCNE2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:100-1:2000; IHC: 1:2500

Type: Primary
Antigen: CCNE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR