Anti-VEPH1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590277EA 625 EUR
ORIGTA590277
Anti-VEPH1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot 1:20000, ELISA 1:100-1:2000

Type: Primary
Antigen: VEPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR