Anti-Gtpbp10 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA329554EA 363 EUR
ORIGTA329554
Anti-Gtpbp10 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Gtpbp10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR