Anti-NFYA Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA329254EA 575 EUR
ORIGTA329254
Anti-NFYA Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NFYA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR