Anti-FOXM1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590329EA 568 EUR
ORIGTA590329
Anti-FOXM1 Rabbit Polyclonal Antibody
Recommended Dilutions: ELISA: 1:100-1:2000; IHC: 1:45; Immunofluorescence: 1:11

Type: Primary
Antigen: FOXM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR