Anti-MS4A7 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590327EA 568 EUR
ORIGTA590327
Anti-MS4A7 Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot 1:20000, IHC 1:150, ELISA 1:100-1:2000

Type: Primary
Antigen: MS4A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR