Anti-SLC44A4 Polyclonal Antibody

Proveedor: OriGene
ORIGTA329498EA 575 EUR
ORIGTA329498
Anti-SLC44A4 Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SLC44A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR