Anti-Pla2g12a Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA329489EA 575 EUR
ORIGTA329489
Anti-Pla2g12a Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Pla2g12a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR