Anti-TFPI Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590219EA 568 EUR
ORIGTA590219
Anti-TFPI Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:100-1:2000; IHC: 1:250; Immunofluorescence: 1:62

Type: Primary
Antigen: TFPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR