Anti-IL17RA Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590245EA 568 EUR
ORIGTA590245
Anti-IL17RA Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:5000-1:20000; ELISA: 1:100-1:2000

Type: Primary
Antigen: IL17RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR