Anti-FSTL1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590234EA 684 EUR
ORIGTA590234
Anti-FSTL1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot 1:5000-20000, ELISA 1:100-1:2000

Type: Primary
Antigen: FSTL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR