Anti-THY1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590220EA 568 EUR
ORIGTA590220
Anti-THY1 Rabbit Polyclonal Antibody
Recommended Dilutions: ELISA: 1:100-1:2000

Type: Primary
Antigen: THY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR