Anti-DEFA1B Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA328408EA 514 EUR
ORIGTA328408
Anti-DEFA1B Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: DEFA1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR