Anti-NOV Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA328406EA 564 EUR
ORIGTA328406
Anti-NOV Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NOV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR