Anti-CCL19 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA328395EA 564 EUR
ORIGTA328395
Anti-CCL19 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CCL19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR