Anti-MIA2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA328384EA 496 EUR
ORIGTA328384
Anti-MIA2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MIA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR