Anti-CCL7 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA328378EA 466 EUR
ORIGTA328378
Anti-CCL7 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CCL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR