Anti-PARK7 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326226EA 514 EUR
ORIGTA326226
Anti-PARK7 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100Immunofluorescence: 1:500-1:1000 ELISA: 1:10000

Type: Primary
Antigen: PARK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR