Anti-CDKN1C Mouse Monoclonal Antibody [clone: KP10]

Proveedor: OriGene
ORIGTA327720EA 1020 EUR
ORIGTA327720
Anti-CDKN1C Mouse Monoclonal Antibody [clone: KP10]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: CDKN1C
Clonality: Monoclonal
Clone: KP10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR