Anti-POU5F1 Mouse Monoclonal Antibody [clone: MRQ-10]

Proveedor: OriGene
ORIGTA327717EA 1790 EUR
ORIGTA327717
Anti-POU5F1 Mouse Monoclonal Antibody [clone: MRQ-10]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: POU5F1
Clonality: Monoclonal
Clone: MRQ-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR