Anti-NEFH Mouse Monoclonal Antibody [clone: 2F11]

Proveedor: OriGene
ORIGTA327713EA 816 EUR
ORIGTA327713
Anti-NEFH Mouse Monoclonal Antibody [clone: 2F11]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: NEFH
Clonality: Monoclonal
Clone: 2F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR