Anti-MYH11 Mouse Monoclonal Antibody [clone: SMMS-1]

Proveedor: OriGene
ORIGTA327710EA 895 EUR
ORIGTA327710
Anti-MYH11 Mouse Monoclonal Antibody [clone: SMMS-1]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: MYH11
Clonality: Monoclonal
Clone: SMMS-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR