Anti-MYOG Mouse Monoclonal Antibody [clone: F5D]

Proveedor: OriGene
ORIGTA327708EA 898 EUR
ORIGTA327708
Anti-MYOG Mouse Monoclonal Antibody [clone: F5D]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: MYOG
Clonality: Monoclonal
Clone: F5D
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR