Anti-MUC6 Mouse Monoclonal Antibody [clone: MRQ-20]

Proveedor: OriGene
ORIGTA327704EA 900 EUR
ORIGTA327704
Anti-MUC6 Mouse Monoclonal Antibody [clone: MRQ-20]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: MUC6
Clonality: Monoclonal
Clone: MRQ-20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR