Anti-MLANA Mouse Monoclonal Antibody [clone: M2-7C10]

Proveedor: OriGene
ORIGTA327699EA 896 EUR
ORIGTA327699
Anti-MLANA Mouse Monoclonal Antibody [clone: M2-7C10]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: M2-7C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR