Anti-CDH16 Mouse Monoclonal Antibody [clone: MRQ-33]

Proveedor: OriGene
ORIGTA327696EA 1080 EUR
ORIGTA327696
Anti-CDH16 Mouse Monoclonal Antibody [clone: MRQ-33]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: CDH16
Clonality: Monoclonal
Clone: MRQ-33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR