Anti-INS Guinea pig Polyclonal Antibody

Proveedor: OriGene
ORIGTA327695EA 813 EUR
ORIGTA327695
Anti-INS Guinea pig Polyclonal Antibody
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: INS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea pig
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR