Anti-GCSAM Mouse Monoclonal Antibody [clone: MRQ-49]

Proveedor: OriGene
ORIGTA327691EA 1270 EUR
ORIGTA327691
Anti-GCSAM Mouse Monoclonal Antibody [clone: MRQ-49]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: GCSAM
Clonality: Monoclonal
Clone: MRQ-49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR