Anti-CGB Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327689EA 515 EUR
ORIGTA327689
Anti-CGB Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:250

Type: Primary
Antigen: CGB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR