Anti-GPC3 Mouse Monoclonal Antibody [clone: 1G12]

Proveedor: OriGene
ORIGTA327687EA 1380 EUR
ORIGTA327687
Anti-GPC3 Mouse Monoclonal Antibody [clone: 1G12]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: GPC3
Clonality: Monoclonal
Clone: 1G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR