Anti-SLC2A1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327686EA 1040 EUR
ORIGTA327686
Anti-SLC2A1 Rabbit Polyclonal Antibody
Anticuerpos
  • Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: SLC2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR