Anti-NOTCH1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326839EA 466 EUR
ORIGTA326839
Anti-NOTCH1 Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: NOTCH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR