Anti-MEF2C Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326189EA 514 EUR
ORIGTA326189
Anti-MEF2C Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000 ELISA: 1:40000

Type: Primary
Antigen: MEF2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR