Anti-GFAP Rabbit Monoclonal Antibody [clone: EP672Y]

Proveedor: OriGene
ORIGTA327682EA 983 EUR
ORIGTA327682
Anti-GFAP Rabbit Monoclonal Antibody [clone: EP672Y]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: EP672Y
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR