Anti-PIP Mouse Monoclonal Antibody [clone: 23A3]

Proveedor: OriGene
ORIGTA327680EA 813 EUR
ORIGTA327680
Anti-PIP Mouse Monoclonal Antibody [clone: 23A3]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: PIP
Clonality: Monoclonal
Clone: 23A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR