Anti-F13A1 Rabbit Monoclonal Antibody [clone: EP3372]

Proveedor: OriGene
ORIGTA327675EA 898 EUR
ORIGTA327675
Anti-F13A1 Rabbit Monoclonal Antibody [clone: EP3372]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: F13A1
Clonality: Monoclonal
Clone: EP3372
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR