Anti-MUC1 Mouse Monoclonal Antibody [clone: E29]

Proveedor: OriGene
ORIGTA327672EA 567 EUR
ORIGTA327672
Anti-MUC1 Mouse Monoclonal Antibody [clone: E29]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: E29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR