Anti-DES Mouse Monoclonal Antibody [clone: D33]

Proveedor: OriGene
ORIGTA327670EA 979 EUR
ORIGTA327670
Anti-DES Mouse Monoclonal Antibody [clone: D33]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: DES
Clonality: Monoclonal
Clone: D33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR