Anti-KRT5 Mouse Monoclonal Antibody [clone: D5/16B4]

Proveedor: OriGene
ORIGTA327667EA 815 EUR
ORIGTA327667
Anti-KRT5 Mouse Monoclonal Antibody [clone: D5/16B4]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: KRT5
Clonality: Monoclonal
Clone: D5/16B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR